Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412961

Sigma-Aldrich

Tetraphenylolethane glycidyl ether

Synonym(s):

1,1,2,2-Tetra(p -hydroxyphenyl)ethane tetraglycidyl ether, 1,1,2,2-Tetrakis(4-glycidoxyphenyl)ethane, 1-phenyleneoxymethylene)]tetrakis[oxirane], 2,2′,2′′,2′′′-[1,2-Ethanediylidenetetrakis(4,1-phenyleneoxymethylene)]tetrakis[oxirane]

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

chips

specific activity

>3 glycidyl/molecule

mp

77-83 °C (lit.)

InChI

1S/C38H38O8/c1-9-29(39-17-33-21-43-33)10-2-25(1)37(26-3-11-30(12-4-26)40-18-34-22-44-34)38(27-5-13-31(14-6-27)41-19-35-23-45-35)28-7-15-32(16-8-28)42-20-36-24-46-36/h1-16,33-38H,17-24H2

InChI key

HDDQXUDCEIMISH-UHFFFAOYSA-N

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kana Nishihara et al.
Mutagenesis, 31(1), 69-81 (2015-08-06)
DNA repair pathways play a critical role in maintaining cellular homeostasis by repairing DNA damage induced by endogenous processes and xenobiotics, including environmental chemicals. Induction of DNA damage may lead to genomic instability, disruption of cellular homeostasis and potentially tumours.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service