Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

411957

Sigma-Aldrich

Pentafluorophenol-OD

98 atom % D

Synonym(s):

Pentafluorophen(ol-d)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5OD
CAS Number:
Molecular Weight:
185.07
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21
assay:
≥99% (GC)
technique(s):
NMR: suitable
bp:
143 °C (lit.)

isotopic purity

98 atom % D

assay

≥99% (GC)

form

solid

technique(s)

NMR: suitable

bp

143 °C (lit.)

mp

34-36 °C (lit.)

mass shift

M+1

SMILES string

[2H]Oc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6HF5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H/i/hD

InChI key

XBNGYFFABRKICK-DYCDLGHISA-N

General description

Pentafluorophen(ol-d) is pentafluorophenol with the hydrogen on –OH substituted by deuterium. It is an NMR solvent.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced effects of lithocholic acid incorporation into liquid-crystalline biopolymer poly (coumaric acid) on structural ordering and cell adhesion.
Matsusaki M, et al.
Biomaterials, 26(32), 6263-6270 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service