Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40842

Sigma-Aldrich

Tris(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)tin azide

≥90%

Synonym(s):

Azidotris(3,3,4,4,5,5,6,6,7,7,8,8,8- tridecafluorooctyl)stannane, Tris(1H,1H,2H,2H-perfluorooctyl)tin azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H12F39N3Sn
CAS Number:
Molecular Weight:
1202.02
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥90%

bp

>280 °C

mp

6 °C

density

1.840 g/mL at 25 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Sn](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)N=[N+]=[N-]

InChI

1S/3C8H4F13.N3.Sn/c3*1-2-3(9,10)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21;1-3-2;/h3*1-2H2;;/q;;;-1;+1

InChI key

XBCIYHZQPNYTIF-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Protected by U.S. patents 6,156,896; 5,859,247; and 5,777,125

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service