Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407976

Sigma-Aldrich

Ethylene glycol dicyclopentenyl ether methacrylate

Synonym(s):

2-(Dicyclopentenyloxy)ethyl methacrylate, Dicyclopentenyloxyethyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O3
CAS Number:
Molecular Weight:
262.34
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

contains

200-600 ppm MEHQ as inhibitor

refractive index

n20/D 1.497 (lit.)

density

1.064 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCOC1CC2CC1C3CC=CC23

InChI

1S/C16H22O4/c1-12(2)16(17)18-11-15(19-13-7-3-4-8-13)20-14-9-5-6-10-14/h7,9,15H,1,3-6,8,10-11H2,2H3

Inchi Key

JMIZWXDKTUGEES-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P K Chan et al.
Journal of toxicology and environmental health, 16(1), 39-54 (1985-01-01)
The subchronic dermal toxicity of dicyclopentenyloxethyl methacrylate (DPOMA) was evaluated in young adult New Zealand White rabbits, and its potential to produce delayed contact sensitization was evaluated in Harley guinea pigs by a modified Buehler's closed patch technique. In addition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service