Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407542

Sigma-Aldrich

Di(ethylene glycol) 2-ethylhexyl ether acrylate

contains 500 ppm monomethyl ether hydroquinone as inhibitor, 95%

Synonym(s):

2-Ethylhexyl carbitol acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CH2OCH2CH2OCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

95%

form

liquid

contains

500 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.447 (lit.)

density

0.946 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COCCOCCOC(=O)C=C

InChI

1S/C15H28O4/c1-4-7-8-14(5-2)13-18-10-9-17-11-12-19-15(16)6-3/h6,14H,3-5,7-13H2,1-2H3

InChI key

XNMJQRPYVCIXGZ-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service