Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407399

Sigma-Aldrich

Tetrakis(diethylamido)titanium(IV)

technical grade

Synonym(s):

TDEAT, Tetrakis(diethylamino)titanium(IV), Titanium(IV) tetrakis(diethylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C2H5)2N]4Ti
CAS Number:
Molecular Weight:
336.38
Beilstein/REAXYS Number:
3621431
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical grade

assay

>95% (perchloric acid titration)

form

liquid

reaction suitability

core: titanium
reagent type: catalyst

refractive index

n20/D 1.536 (lit.)

bp

112 °C/0.1 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)[Ti](N(CC)CC)(N(CC)CC)N(CC)CC

InChI

1S/4C4H10N.Ti/c4*1-3-5-4-2;/h4*3-4H2,1-2H3;/q4*-1;+4

InChI key

VJDVOZLYDLHLSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

-19.8 °F - closed cup

flash_point_c

-28.8 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service