Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

406848

Sigma-Aldrich

Tetraisobutyldialuminoxane solution

10 wt. % in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlOAl[CH2CH(CH3)2]2
CAS Number:
Molecular Weight:
298.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

10 wt. % in toluene

bp

111 °C

density

0.867 g/mL at 25 °C

SMILES string

CC(C)C[Al](CC(C)C)O[Al](CC(C)C)CC(C)C

InChI

1S/4C4H9.2Al.O/c4*1-4(2)3;;;/h4*4H,1H2,2-3H3;;;

Inchi Key

CTNFGBKAHAEKFE-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

17.6 °F - closed cup

flash_point_c

-8 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service