406627
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol) bis(2-aminopropyl ether)
average Mn ~900
Synonym(s):
Jeffamine® ED-900
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
feed ratio
3.5:15.5 (PO:EO)
mol wt
average Mn ~900
refractive index
n20/D 1.4657
bp
240 °C (lit.)
mp
22 °C
density
1.056 g/mL at 25 °C
SMILES string
OCCO.CC(N)CO.CC(O)CO
Looking for similar products? Visit Product Comparison Guide
Legal Information
Jeffamine is a registered trademark of Huntsman Petrochemical, LLC
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service