Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

405787

Sigma-Aldrich

Dimesitylmethane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[2,4,6-(CH3)3C6H2]2CH2
CAS Number:
Molecular Weight:
252.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

solid

mp

132-135 °C (lit.)

SMILES string

Cc1cc(C)c(Cc2c(C)cc(C)cc2C)c(C)c1

InChI

1S/C19H24/c1-12-7-14(3)18(15(4)8-12)11-19-16(5)9-13(2)10-17(19)6/h7-10H,11H2,1-6H3

InChI key

ZQYQPPLKPFBKLD-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formation of seven-membered carbocycles by the use of cyclopropyl silyl ethers as homoenols.
Oleg L Epstein et al.
Angewandte Chemie (International ed. in English), 45(30), 4988-4991 (2006-07-05)
Fangwei Shao et al.
Bioconjugate chemistry, 19(12), 2487-2491 (2008-12-05)
A facile synthetic route to prepare monofunctional carbocyanine dyes for biological application is developed. Three pentamethine carbocyanine dyes have been successfully modified with a variety of functional groups such as: carboxylic acids, azides, or alkynes. The new dyes are characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service