Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40308

Sigma-Aldrich

Acetone

Synonym(s):

Dimethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein/REAXYS Number:
635680
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021201
PubChem Substance ID:
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

form

liquid

CofA

specification on request

expl. lim.

13.2 %

color

clear colorless

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
Semiconductor grade acetone may be used for cleaning of the surfaces, such as;
  • Hydrogen terminated silicon wafer.[1][2]
  • SiC wafers.[3]
  • Chromium nitride epilayer on silicon wafer.[4]

The semiconductor grade acetone was also used in a comparative study for observing the effect of various solvents on nitrogen containing promoters (NCPs), which is used in oxidation reaction of 5-hydroxymethylfurfural.[5]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cu catalyzed oxidation of 5-hydroxymethylfurfural to 2, 5-diformylfuran and 2, 5-furandicarboxylic acid under benign reaction conditions.
Hansen T, et al.
Applied Catalysis A: General, 456, 44-50 (2013)
Organic monolayers from 1-alkynes covalently attached to chromium nitride: alkyl and fluoroalkyl termination.
Pujari S, et al.
Langmuir, 29(33), 10393-10404 (2013)
Hexadecadienyl monolayers on hydrogen-terminated Si (111): faster monolayer formation and improved surface coverage using the enyne moiety
Rijksen B, et al.
Langmuir, 28(16), 6577-6588 (2012)
Covalently attached organic monolayers onto silicon carbide from 1-alkynes: molecular structure and tribological properties.
Pujari S, et al.
Langmuir, 29(13), 4019-4031 (2013)
Mono-fluorinated alkyne-derived SAMs on oxide-free Si (111) surfaces: preparation, characterization and tuning of the Si workfunction
Pujari S, et al.
Langmuir, 29(2), 570-580 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service