Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40307

Sigma-Aldrich

Hydrogen peroxide solution

≥30%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein/REAXYS Number:
3587191
MDL number:
UNSPSC Code:
12352304
PubChem Substance ID:

vapor pressure

23.3 mmHg ( 30 °C)

assay

≥30%

CofA

specification on request

shelf life

~2 yr

reaction suitability

reagent type: oxidant

concentration

25-35%

density

1.110 g/cm3

storage temp.

2-8°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide is a strong oxidizer. It may be produced industrially by anthraquinone oxidation.[1] it is used in semiconductor industry essentially for cleaning silicon wafers, removal of photoresists and to etch metallic copper on printed circuit boards. Ultrapurification of hydrogen peroxide by reverse osmosis has been reported in detail. [2]

Application

Semiconductor grade hydrogen peroxide was used to etch TiN[3] 2 and GaAs structures. [4]

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TiN metal hardmask etch residue removal on advanced porous low-k and Cu device with corner rounding scheme
Cui H, et al.
Solid State Phenomena, 187 (2012)
Localized GaAs etching with acidic hydrogen peroxide solutions
Shaw DW, et al.
Journal of the Electrochemical Society, 128(4), 874-880 (1981)
Ultrapurification of hydrogen peroxide solution from ionic metals impurities to semiconductor grade by reverse osmosis.
Abejon R, et al.
Separation and Purification Technology, 76(1), 44-51 (2010)
Jose M Campos-Martin et al.
Angewandte Chemie (International ed. in English), 45(42), 6962-6984 (2006-10-14)
Hydrogen peroxide (H2O2) is widely used in almost all industrial areas, particularly in the chemical industry and environmental protection. The only degradation product of its use is water, and thus it has played a large role in environmentally friendly methods
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service