402729
2-Methyl-5-nitroaniline
95%
Synonym(s):
2-Amino-4-nitrotoluene, 5-Nitro-o-toluidine
About This Item
Recommended Products
assay
95%
form
powder
mp
103-106 °C (lit.)
functional group
nitro
SMILES string
Cc1ccc(cc1N)[N+]([O-])=O
InChI
1S/C7H8N2O2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,8H2,1H3
Inchi Key
DSBIJCMXAIKKKI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service