Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

401498

Sigma-Aldrich

Zinc hexafluorosilicate hydrate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSiF6 · xH2O
Molecular Weight:
207.47 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99%
form:
solid

assay

99%

form

solid

SMILES string

O.F[Zn]F.F[Si](F)(F)F

InChI

1S/F4Si.2FH.H2O.Zn/c1-5(2,3)4;;;;/h;2*1H;1H2;/q;;;;+2/p-2

InChI key

GUJNEJDSEHACGC-UHFFFAOYSA-L

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service