Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

401358

Sigma-Aldrich

(S)-(−)-N,α-Dimethylbenzylamine

98%

Synonym(s):

(S)-DMBA, (S)-(−)-N-Methyl-1-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCH3
CAS Number:
Molecular Weight:
135.21
Beilstein/REAXYS Number:
1446867
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

optical activity

[α]20/D −73±5°, c = 2 in chloroform

refractive index

n20/D 1.512 (lit.)

bp

74-76 °C/11 mmHg (lit.)

density

0.928 g/mL at 20 °C (lit.)

functional group

amine
phenyl

SMILES string

CN[C@@H](C)c1ccccc1

InChI

1S/C9H13N/c1-8(10-2)9-6-4-3-5-7-9/h3-8,10H,1-2H3/t8-/m0/s1

InChI key

RCSSHZGQHHEHPZ-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-N,a-Dimethylbenzylamine may be used along with benzaldehyde in the Mannich-type aminoalkylation of 2-naphthol to form 1-((S)-phenyl(((1′S)-1′-phenylethyl)methylamino)methyl)-2-naphthol. This optically active tertiary aminonaphthol can catalyze the enantioselective alkenylation of various aldehyde to generate chiral (E)- allyl alcohols.[1]

Application

Used to make asymmetric organometallic catalysts (e.g., for conjugate additions to enones).[2]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A convenient, one-step synthesis of optically active tertiary aminonaphthol and its applications in the highly enantioselective alkenylations of aldehydes
J JX, et al.
The Journal of Organic Chemistry, 68(4), 1589-1590 (2003)
Tetrahedron Letters, 31, 965-965 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service