Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3DXPLA011

Sigma-Aldrich

PLA 3D printing filament

greener alternative

red, diam. 1.75 mm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

1.75 mm

color

red

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

3DXTech is made in the USA using 100% virgin resin and colorants - precision extruded onto 1kg reels (2.2 lbs.). The filament is then vacuum-sealed with desiccant to protect it from moisture.  
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry.This product belongs to enabling category of greener alternatives and aligns with "Design for energy efficiency". PLA filament is one of the easiest material to 3D print with. Its an eco-friendly material, as its biodegradable. PLA filament is non-toxic and requies less energy to 3D print. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.

Preparation Note

Recommended Print Conditions: Extruder Temp: 190-220°C. Bed Temp: 23-60°C [No heat bed required]

Legal Information

Product of 3DXTech
3DXTech is a trademark of 3DXTech

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Three-dimensional printing technology applications range from personal tools to aerospace equipment.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service