3DXCFR003
CarbonX™ CFR-ABS carbon fiber reinforced ABS 3D printing filament
black, diam. 1.75 mm, weight 750 gper each reel
About This Item
Recommended Products
diam.
1.75 mm
weight
750 gper each reel
color
black
InChI
1S/C8H8.C4H6.C3H3N/c1-2-8-6-4-3-5-7-8;1-3-4-2;1-2-3-4/h2-7H,1H2;3-4H,1-2H2;2H,1H2
InChI key
XECAHXYUAAWDEL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
- 3687 MPa Flexural Modulus (85% improvement over unfilled ABS)
- 3458 MPa Tensile Modulus (77% improvement over unfilled ABS)
Preparation Note
- Extruder: 220 - 235 °C. CarbonX™ CFR is a filled product and has a higher melt viscosity vs. unfilled resin. Therefore; it is sometimes necessary to print at higher temperatures to allow the resin to flow properly.
- Bed Temp: 100-110 °C
- Nozzle: minimum 0.35 mm orifice recommended
- Carbon fiber filamenet is more abrasive than unfilled materials; so extended use will results in more wear on your nozzle.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.
Three-dimensional printing technology applications range from personal tools to aerospace equipment.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service