Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3DXCFR001

Sigma-Aldrich

CarbonX CFR-ABS carbon fiber reinforced ABS 3D printing filament

black, diam. 2.85 mm, weight 750 gper each reel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

diam.

2.85 mm

weight

750 gper each reel

color

black

InChI

1S/C8H8.C4H6.C3H3N/c1-2-8-6-4-3-5-7-8;1-3-4-2;1-2-3-4/h2-7H,1H2;3-4H,1-2H2;2H,1H2

InChI key

XECAHXYUAAWDEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CarbonX CFR-ABS is a high-performance carbon fiber reinforced ABS filament ideal for anyone that desires a structural component with high modulus, excellent surface quality, light weight, and ease of printing. Made using premium Sabic MG-94 ABS and high modulus carbon fiber.

  • 3687 MPa Flexural Modulus (85% improvement over unfilled ABS)
  • 3458 MPa Tensile Modulus (77% improvement over unfilled ABS)

Preparation Note

Recommended Print Conditions:

  • Extruder: 220 - 235 °C. CarbonX CFR is a filled product and has a higher melt viscosity vs. unfilled resin. Therefore; it is sometimes necessary to print at higher temperatures to allow the resin to flow properly.
  • Bed Temp: 100-110 °C
  • Nozzle: minimum 0.35 mm orifice recommended
  • Carbon fiber filamenet is more abrasive than unfilled materials; so extended use will results in more wear on your nozzle.

Legal Information

CarbonX is a trademark of 3DXTech

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Three-dimensional printing technology applications range from personal tools to aerospace equipment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service