Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3DXABS003

Sigma-Aldrich

ABS 3D printing filament

greener alternative

blue, diam. 1.75 mm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

1.75 mm

color

blue

greener alternative category

InChI

1S/C8H8.C4H6.C3H3N/c1-2-8-6-4-3-5-7-8;1-3-4-2;1-2-3-4/h2-7H,1H2;3-4H,1-2H2;2H,1H2

InChI key

XECAHXYUAAWDEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3DXTech ABS is made in the USA using 100% virgin resin and colorants - precision extruded onto 1kg reels (2.2 lbs.). The filament is then vacuum-sealed with desiccant to protect it from moisture.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and thus aligns with "Design for energy efficiency". ABS 3D printing filament is known to be strong and durable material. It is resistant to heat and everyday chemicals. It can also endure more to stress, heat and pressure as compared to other 3D printing materials. Click here for more information.

Application

This product can be used in material extrusion 3D printing technique.

Preparation Note

Recommended Print Conditions: Extruder Temp: 220-240°C, ideal 230°C. Bed Temp:100-110°C. Bed Preparation: Polyimide Tape, ABS/Acetone Slurry

Legal Information

Product of 3DXTech
3DXTech is a trademark of 3DXTech

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Three-dimensional printing technology applications range from personal tools to aerospace equipment.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service