3DXABS002
ABS 3D printing filament
black, diam. 2.85 mm
About This Item
Recommended Products
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
diam.
2.85 mm
color
black
greener alternative category
, Enabling
InChI
1S/C8H8.C4H6.C3H3N/c1-2-8-6-4-3-5-7-8;1-3-4-2;1-2-3-4/h2-7H,1H2;3-4H,1-2H2;2H,1H2
InChI key
XECAHXYUAAWDEL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Preparation Note
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Three-dimensional printing technology applications range from personal tools to aerospace equipment.
Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service