Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
AlB12
CAS Number:
Molecular Weight:
156.71
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
234-924-2
MDL number:
Skip To
SMILES string
[Al].B1=BB=BB=BB=BB=BB=B1
form
powder
particle size
−325 mesh
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service