Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39897

Sigma-Aldrich

Chloro-methoxy-dimethylsilane

technical, ~85% (GC)

Synonym(s):

Chloro-dimethyl-methoxysilane, Dimethyl-chloromethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9ClOSi
CAS Number:
Molecular Weight:
124.64
Beilstein/REAXYS Number:
2035892
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

technical

assay

~85% (GC)

SMILES string

CO[Si](C)(C)Cl

InChI

1S/C3H9ClOSi/c1-5-6(2,3)4/h1-3H3

InChI key

AWSNMQUTBVZKHY-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

17.6 °F - closed cup

flash_point_c

-8 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service