Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39793

Sigma-Aldrich

Boc-(R)-α-(4-tert-butylbenzyl)-Pro-OH

≥97.0% (HPLC)

Synonym(s):

(R)-1-Boc-2-(4-tert-butylbenzyl)-2-pyrrolidinecarboxylic acid, Boc-α-(4-tert-butylbenzyl)-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31NO4
CAS Number:
Molecular Weight:
361.48
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Product 39793 is not currently sold in your country. Contact Technical Service

Quality Level

assay

≥97.0% (HPLC)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCC[C@@]1(Cc2ccc(cc2)C(C)(C)C)C(O)=O

InChI

1S/C21H31NO4/c1-19(2,3)16-10-8-15(9-11-16)14-21(17(23)24)12-7-13-22(21)18(25)26-20(4,5)6/h8-11H,7,12-14H2,1-6H3,(H,23,24)/t21-/m1/s1

InChI key

BOEGKJLPJARVIT-OAQYLSRUSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service