396478
2-Pyridyl trifluoromethanesulfonate
98%
Synonym(s):
2-Pyridyl triflate
About This Item
Recommended Products
Quality Level
assay
98%
form
liquid
refractive index
n20/D 1.435 (lit.)
bp
108 °C/25 mmHg (lit.)
density
1.477 g/mL at 25 °C (lit.)
functional group
fluoro
triflate
SMILES string
FC(F)(F)S(=O)(=O)Oc1ccccn1
InChI
1S/C6H4F3NO3S/c7-6(8,9)14(11,12)13-5-3-1-2-4-10-5/h1-4H
InChI key
COLRMVLTWJTLFJ-UHFFFAOYSA-N
General description
application
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
80.6 °F - closed cup
flash_point_c
27 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service