394424
(Allyloxycarbonylmethyl)triphenylphosphonium iodide
97%
About This Item
Recommended Products
assay
97%
reaction suitability
reaction type: C-C Bond Formation
mp
138 °C (dec.) (lit.)
SMILES string
[I-].C=CCOC(=O)C[P+](c1ccccc1)(c2ccccc2)c3ccccc3
InChI
1S/C23H22O2P.HI/c1-2-18-25-23(24)19-26(20-12-6-3-7-13-20,21-14-8-4-9-15-21)22-16-10-5-11-17-22;/h2-17H,1,18-19H2;1H/q+1;/p-1
InChI key
PJGRTFGJXKYEIH-UHFFFAOYSA-M
Application
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service