Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

393126

Sigma-Aldrich

Fast Red B Salt

Dye content 20 %

Synonym(s):

2-Methoxy-4-nitrobenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 5, Azoic diazo component 5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8Cl3N3O3Zn
CAS Number:
Molecular Weight:
353.91
Colour Index Number:
37125
UNSPSC Code:
41116121
PubChem Substance ID:

form

powder

composition

Dye content, 20%

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

InChI

1S/C7H7N3O3.3ClH.Zn/c1-13-7-4-5(10(11)12)2-3-6(7)9-8;;;;/h2-4,8H,1H3;3*1H;/q;;;;+2/p-2

InChI key

GXOUZINWMJUARK-UHFFFAOYSA-L


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian R Fritzsche et al.
BMC cancer, 8, 381-381 (2008-12-23)
Enhanced activity of histone deacetylases (HDAC) is associated with more aggressive tumour behaviour and tumour progression in various solid tumours. The over-expression of these proteins and their known functions in malignant neoplasms has led to the development of HDAC inhibitors
V Shalhoub et al.
Calcified tissue international, 89(2), 140-150 (2011-06-03)
Elevated serum levels of the phosphate-regulating hormone fibroblast growth factor 23 (FGF23) are found in patients with phosphate wasting diseases and chronic kidney disease-mineral and bone disorder (CKD-MBD). These diseases are associated with rickets and renal osteodystrophy, respectively. FGF23 is
Jutta Schade et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(2), 147-155 (2007-10-31)
The expression of dipeptidyl peptidase 4 (DP4, CD26) affects T-cell recruitment to lungs in an experimental rat asthma model. Furthermore, the gene of the structural homologous DP10 represents a susceptibility locus for asthma in humans, and the functional homologous DP8/9

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service