About This Item
assay
98%
form
liquid
refractive index
n20/D 1.429 (lit.)
bp
46 °C/10 mmHg (lit.)
density
1.019 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
COC1OC(C)(OC)C=C1
InChI
1S/C7H12O3/c1-7(9-3)5-4-6(8-2)10-7/h4-6H,1-3H3
InChI key
ODARVONGEVVSAG-UHFFFAOYSA-N
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service