Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

392464

Sigma-Aldrich

4,5-Diamino-6-hydroxy-2-mercaptopyrimidine hemisulfate salt hydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4OS · 0.5H2SO4 · xH2O
CAS Number:
Molecular Weight:
207.22 (anhydrous basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

235 °C (dec.) (lit.)

SMILES string

[H]O[H].OS(O)(=O)=O.Nc1nc(S)nc(O)c1N

InChI

1S/C4H6N4OS.H2O4S.H2O/c5-1-2(6)7-4(10)8-3(1)9;1-5(2,3)4;/h5H2,(H4,6,7,8,9,10);(H2,1,2,3,4);1H2

InChI key

FPNMSAFUGFYKCX-UHFFFAOYSA-N

General description

4,5-Diamino-6-hydroxy-2-mercaptopyrimidine hemisulfate salt hydrate is a pyrimidine derivative. IR spectral investigations of 4,5-diamino-6-hydroxy-2-mercaptopyrimidine have been reported.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society: Faraday transactions. Molecular and chemical physics. II, 82(1-4), 123-125 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service