392324
2-(Dibenzylamino)-4′-hydroxy-3′-methoxyacetophenone hydrochloride
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H3(OCH3)COCH2N(CH2C6H5)2 · HCl
CAS Number:
Molecular Weight:
397.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
form
powder
mp
200 °C (dec.) (lit.)
SMILES string
Cl.COc1cc(ccc1O)C(=O)CN(Cc2ccccc2)Cc3ccccc3
InChI
1S/C23H23NO3.ClH/c1-27-23-14-20(12-13-21(23)25)22(26)17-24(15-18-8-4-2-5-9-18)16-19-10-6-3-7-11-19;/h2-14,25H,15-17H2,1H3;1H
InChI key
HEVRTBQBWPPTET-UHFFFAOYSA-N
General description
2-(Dibenzylamino)-4′-hydroxy-3′-methoxyacetophenone hydrochloride is an acetophenone derivative.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service