Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

391700

Sigma-Aldrich

Croconic acid

new

98%

Synonym(s):

4,5-Dihydroxy-4-cyclopentene-1,2,3-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2O5
CAS Number:
Molecular Weight:
142.07
Beilstein/REAXYS Number:
2043130
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

98%

form

powder, crystals or chunks

mp

>300 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC1=C(O)C(=O)C(=O)C1=O

InChI

1S/C5H2O5/c6-1-2(7)4(9)5(10)3(1)8/h6-7H

InChI key

RBSLJAJQOVYTRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoting Gao et al.
Biomaterials, 267, 120454-120454 (2020-11-08)
The development of tumor acidic microenvironment-responsive theranostic agents is a research hotspot. Herein, we developed highly photostable amphiphilic croconium dye-anchored red blood cell membrane vesicle (denoted as LET-5) for tumor pH-responsive near-infrared fluorescence (NIRF) and photoacoustic (PA) duplex imaging-guided photothermal
Francisco Colmenero et al.
The journal of physical chemistry. A, 123(19), 4241-4261 (2019-04-26)
A precise and complete thermodynamic, Raman spectroscopic, and ultraviolet-visible (UV-vis) optical characterization of the deltic, squaric, and croconic cyclic oxocarbon acids is obtained using theoretical solid-state methods employing very demanding calculation parameters. The computed fundamental thermodynamic properties include the isobaric
Lan Yao et al.
Polymers, 12(5) (2020-05-28)
Polymeric nanomaterials made from amphiphilic block copolymers are increasingly used in the treatment of tumor tissues. In this work, we firstly synthesized the amphiphilic block copolymer PBnMA-b-P(BAPMA-co-PEGMA) via reversible addition-fragmentation chain transfer (RAFT) polymerization using benzyl methacrylate (BnMA), poly (ethylene
Felicitas Jansen et al.
Macromolecular rapid communications, 41(22), e2000418-e2000418 (2020-10-14)
In the development and optimization of imaging methods, photoacoustic imaging (PAI) has become a powerful tool for preclinical biomedical diagnosis and detection of cancer. PAI probes can improve contrast and help identify pathogenic tissue. Such contrast agents must meet several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service