Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39166

Sigma-Aldrich

Boc-(S)-α-(4-tert-butylbenzyl)-Pro-OH

purum, ≥98.0% (HPLC)

Synonym(s):

(S)-1-Boc-2-(4-tert-butylbenzyl)-2-pyrrolidinecarboxylic acid, Boc-α-(4-tert-butylbenzyl)-D-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31NO4
CAS Number:
Molecular Weight:
361.48
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

grade

purum

assay

≥98.0% (HPLC)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1CCC[C@]1(Cc2ccc(cc2)C(C)(C)C)C(O)=O

InChI

1S/C21H31NO4/c1-19(2,3)16-10-8-15(9-11-16)14-21(17(23)24)12-7-13-22(21)18(25)26-20(4,5)6/h8-11H,7,12-14H2,1-6H3,(H,23,24)/t21-/m0/s1

Inchi Key

BOEGKJLPJARVIT-NRFANRHFSA-N

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen M Rodgers et al.
Pharmacology, biochemistry, and behavior, 194, 172935-172935 (2020-04-27)
The long-term treatment of chronic pain by opioids is limited by tolerance and risk of addiction/dependence. Previously, we have shown that combination treatment of morphine with a dopamine D1 or D3 receptor modulator restored morphine analgesia in morphine-resistant neuropathic pain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service