All Photos(1)
About This Item
Linear Formula:
(CD3)3P
CAS Number:
Molecular Weight:
85.13
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
vapor pressure
7.73 psi ( 20 °C)
isotopic purity
99 atom % D
bp
38 °C (lit.)
density
0.821 g/mL at 25 °C (lit.)
mass shift
M+9
Storage temp.
2-8°C
SMILES string
[2H]C([2H])([2H])P(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C3H9P/c1-4(2)3/h1-3H3/i1D3,2D3,3D3
Inchi Key
YWWDBCBWQNCYNR-GQALSZNTSA-N
Application
Has been employed in studies of organometallic compounds.
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
accessory
Product No.
Description
Pricing
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
-2.2 °F - closed cup
flash_point_c
-19.00 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of Organometallic Chemistry, 428, 99-99 (1992)
Organometallics, 10, 1577-1577 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service