38943
1-(3-Cyanopropyl)-3-methylimidazolium bis(trifluoromethylsulfonyl)amide
Task Specific Ionic Liquid (TSIL), ≥98.5% (HPLC/T)
Synonym(s):
1-Methyl-3-(3-cyanopropyl)imidazolium bis(trifluoromethylsulfonyl)amide
About This Item
assay
≥98.5% (HPLC/T)
form
liquid
greener alternative product characteristics
Catalysis
Design for Degradation
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
impurities
≤0.1% water
≤10 mg/kg halogenes (as chloride)
refractive index
n20/D 1.441
greener alternative category
, Aligned
SMILES string
C[n+]1ccn(CCCC#N)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
InChI
1S/C8H12N3.C2F6NO4S2/c1-10-6-7-11(8-10)5-3-2-4-9;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h6-8H,2-3,5H2,1H3;/q+1;-1
InChI key
NNNHNBMBLGKXSW-UHFFFAOYSA-N
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Part III. The first readily biodegradable ionic liquids
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service