Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

389080

Sigma-Aldrich

α-D-Ribofuranose 1,3,5-tribenzoate

97%

Synonym(s):

1,3,5-Tri-O-benzoyl-α-D-ribofuranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O8
CAS Number:
Molecular Weight:
462.45
Beilstein/REAXYS Number:
98446
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

optical activity

[α]20/D +85.0°, c = 1 in chloroform

mp

125-129 °C (lit.)

SMILES string

O=C(OC[C@H]1O[C@H](OC(=O)c2ccccc2)[C@H](OC(=O)c3ccccc3)[C@@H]1OC(=O)c4ccccc4)c5ccccc5

InChI

1S/C26H22O8/c27-21-22(33-24(29)18-12-6-2-7-13-18)20(16-31-23(28)17-10-4-1-5-11-17)32-26(21)34-25(30)19-14-8-3-9-15-19/h1-15,20-22,26-27H,16H2/t20-,21-,22-,26-/m1/s1

InChI key

HUHVPBKTTFVAQF-PIXQIBFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for nucleosides.[1][2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed I Elzagheid et al.
Nucleosides, nucleotides & nucleic acids, 22(5-8), 1339-1342 (2003-10-21)
Interesting and very promising antisense properties of 2'-deoxy-2'-fluoroarabinonucleic acids ((a) Wilds, C.J.; Damha, M.J. 2'-Deoxy-2'-fluoroarabinonucleosides and oligonucleotides (2'F-ANA): synthesis and physicochemical studies. Nucl. Acids Res. 2000, 28, 3625-3635; (b) Viazovkina, E.; Mangos, M.; Elzagheid, M.I.; Damha, M.J. Current Protocols in
Nucleosides, 7, 155-155 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service