Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38899

Sigma-Aldrich

1-Butyl-3-methylimidazolium chloride

≥98.0% (HPLC)

Synonym(s):

BMIMCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15ClN2
CAS Number:
Molecular Weight:
174.67
Beilstein/REAXYS Number:
6449277
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

≥98.0% (HPLC)

impurities

≤0.5% water

mp

~70 °C

SMILES string

[Cl-].CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.ClH/c1-3-4-5-10-7-6-9(2)8-10;/h6-8H,3-5H2,1-2H3;1H/q+1;/p-1

InChI key

FHDQNOXQSTVAIC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium chloride is an imidazolium based ionic liquid.[1]

Application

1-Butyl-3-methylimidazolium chloride can be used as a precursor for the preparation of other ionic liquids by anion metathesis.[2]

Packaging

1kg in plastic bottle
100g in plastic bottle

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

377.6 °F - (External MSDS)

flash_point_c

192 °C - (External MSDS)

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BASIONICS?BASF?s Portfolio of Ionic Liquids
Aldrich Chemfiles, 5(6) null
Effect of ionic liquids, 1-butyl-3-methyl imidazolium bromide and 1-hexyl-3-methyl imidazolium bromide on the vapour-Liquid equilibria of the aqueous d-fructose solutions at 298.15 K and atmospheric pressure using isopiestic method.
Zafarani-Moattar MT, et al.
The Journal of Chemical Thermodynamics, 105, 142-150 (2017)
Muhammad Abdul Haq et al.
Carbohydrate polymers, 223, 115058-115058 (2019-08-21)
It is still challenging to melt-process cellulose. In this paper, we proposed a method for the formation of thermally processable flexible cellulose films via gelation from its solution in ionic liquid (1-butyl-3-methylimidazolium chloride; BMIMCl). Cotton, as a source of cellulose
Fernando Campanhã Vicentini et al.
The Analyst, 139(16), 3961-3967 (2014-06-13)
Voltammetric studies have been carried out using a glassy carbon electrode (GCE) modified with multi-walled carbon nanotubes (MWCNTs) and the ionic liquid 1-butyl-3-methylimidazolium chloride (IL). Studies on the electrochemical properties of GCEs modified with MWCNTs and IL within different polymeric
Q B Zhang et al.
Physical chemistry chemical physics : PCCP, 17(6), 4701-4707 (2015-01-16)
Nanostructured lanthanum was electrochemically prepared on a platinum (Pt) substrate in the room temperature ionic liquid 1-butyl-3-methylimidazolium dicyanamide (BMI-DCA) containing anhydrous LaCl3 at 333 K. The electrochemical reduction behavior of La(iii) was investigated using cyclic voltammetry and chronoamperometry techniques. Cyclic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service