Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

388432

Sigma-Aldrich

2-Aminomethyl-18-crown-6

95%

Synonym(s):

1,4,7,10,13,16-Hexaoxacyclooctadecane-2-methanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H27NO6
CAS Number:
Molecular Weight:
293.36
Beilstein/REAXYS Number:
4184588
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.479 (lit.)

bp

135 °C/0.08 mmHg (lit.)

density

1.126 g/mL at 25 °C (lit.)

SMILES string

NCC1COCCOCCOCCOCCOCCO1

InChI

1S/C13H27NO6/c14-11-13-12-19-8-7-17-4-3-15-1-2-16-5-6-18-9-10-20-13/h13H,1-12,14H2

InChI key

BJUGHLWDFGGTGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminomethyl-18-crown-6 (AM-18-C-6) can be used to prepare:
  • AM-18-C-6 bonded Merrifield peptide resin, which is used to study the absorption and the isotopic effect of magnesium.[1]
  • Schiff base of gossypol with aminomethyl crown ether for complexation of monovalent cations.[2]
  • Crown tagged triarylphosphine by reacting with triphenylphosphine carboxylic acid. These tagged triarylphosphine reagents are used in Mitsunobu and Heck reactions for the removal of phosphine derived by-products.[3]
  • 1,16-hexadecanoyl-bis-(2-aminomethyl)-18-crown-6 (bola A-16), which is utilized as a vesicular carrier in a drug delivery system.[4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new crown ether as vesicular carrier for 5-fluoruracil: synthesis, characterization and drug delivery evaluation
Muzzalupo R, et al.
Colloids and Surfaces. B, Biointerfaces, 58(2), 197-202 (2007)
Synthesis and application of crown ether tagged triarylphosphines
Jackson, Toby and Routledge, Anne
Tetrahedron Letters, 44(6), 1305-1307 (2003)
The Schiff base of gossypol with 2-(aminomethyl)-18-crown-6 complexes and H+, Li+, Na+, K+, Rb+, Cs+ cations studied by ESI MS, 1H NMR, FT-IR and PM5 semiempirical methods
Przybylski P, et al.
Journal of Molecular Structure, 699(1-3), 65-77 (2004)
Adsorption and isotope effects by ion exchange with aqueous-2-aminomethyl-18-crown-6 bonded Merrifield resin
Kim DW, et al.
Journal of Colloid and Interface Science, 254(1), 190-194 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service