Key Documents
388262
Sigma-Aldrich
Poly(vinyl chloride)
average Mn ~67,000, average Mw ~129,000
About This Item
UNSPSC Code:
12352101
Pricing and availability is not currently available.
mol wt
average Mn ~67,000
average Mw ~129,000
inherent viscosity
1.06(lit.)
Warning
May contain vinyl chloride, an OSHA carcinogen. See 29 CFR part 1910
Choose from one of the most recent versions:
Certificates of Analysis (COA)