Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

386278

Sigma-Aldrich

2-tert-Butylfuran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
CAS Number:
Molecular Weight:
124.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

119-120 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccco1

InChI

1S/C8H12O/c1-8(2,3)7-5-4-6-9-7/h4-6H,1-3H3

InChI key

BGQBONSBNSNYIO-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

44.6 °F - closed cup

flash_point_c

7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service