Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

386162

Sigma-Aldrich

2-[3-(3-Ethyl-2(3H)-benzothiazolylidene)-2-methyl-1-propenyl]-3-[3-(sulfooxy)butyl]benzothiazolium hydroxide inner salt

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O4S3
CAS Number:
Molecular Weight:
502.67
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

form

solid

composition

Dye content, 90%

mp

240 °C (dec.) (lit.)

solubility

water and methanol: 1%

λmax

540 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC[n+]1c(\C=C(C)\C=C2\Sc3ccccc3N2CCC(C)S([O-])(=O)=O)sc4ccccc14

InChI

1S/C24H26N2O3S3/c1-4-25-19-9-5-7-11-21(19)30-23(25)15-17(2)16-24-26(14-13-18(3)32(27,28)29)20-10-6-8-12-22(20)31-24/h5-12,15-16,18H,4,13-14H2,1-3H3

InChI key

HHGWLRNRKVHQRS-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service