Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

385441

Sigma-Aldrich

Octyl-functionalized silica gel

phase C8, octyl, matrix irregular silica particle platform, 40-63 μm particle size

Synonym(s):

C8 silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

solid

surface area

550 m2/g

matrix

irregular silica particle platform

matrix active group

C8 phase, octyl

particle size

40-63 μm

pore size

60 Å pore size

Looking for similar products? Visit Product Comparison Guide

Application

For low- and medium-pressure reversed-phase liquid chromatography.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service