Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38456

Sigma-Aldrich

4-(4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecyloxy) benzaldehyde

≥97.0%

Synonym(s):

4-(1H,1H,2H,2H,3H,3H-Perfluoroundecyloxy)benzaldehyde, 4-[3-(Perfluorooctyl)-1-propyloxy]benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11F17O2
CAS Number:
Molecular Weight:
582.25
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥97.0%

form

solid

mp

60-64 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCOc1ccc(C=O)cc1

InChI

1S/C18H11F17O2/c19-11(20,6-1-7-37-10-4-2-9(8-36)3-5-10)12(21,22)13(23,24)14(25,26)15(27,28)16(29,30)17(31,32)18(33,34)35/h2-5,8H,1,6-7H2

InChI key

KOLANEVLEDPTQG-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service