Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

384402

Sigma-Aldrich

Dichloroisopropylphosphine

97%

Synonym(s):

1-Methylethyl)phosphonous dichloride, Isopropyldichlorophosphine, Isopropylphosphonous dichloride, Isopropylphosphorus dichloride, Phosphonous dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHPCl2
CAS Number:
Molecular Weight:
144.97
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.489 (lit.)

density

1.176 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CC(C)P(Cl)Cl

InChI

1S/C3H7Cl2P/c1-3(2)6(4)5/h3H,1-2H3

InChI key

LSMMTQDEKKQXAG-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

113.0 °F - closed cup

flash_point_c

45 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service