Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

382299

Sigma-Aldrich

Silver-exchanged zeolite

pellets, 1.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag84Na2[(AlO2)86(SiO2)106] · xH2O
MDL number:
UNSPSC Code:
12352300

form

pellets

composition

Ag, 9-15%

reaction suitability

core: silver
reagent type: catalyst

particle size

1.6 mm

mp

>500 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Self-binding with high acid stability mordenite crystalline structure efficiently traps halogens from gas streams.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service