Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

381063

Sigma-Aldrich

2,4-Dinitrobenzenesulfonic acid dihydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2O7S · 2H2O
CAS Number:
Molecular Weight:
284.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

98%

form

solid

SMILES string

O.O.OS(=O)(=O)c1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O7S.2H2O/c9-7(10)4-1-2-6(16(13,14)15)5(3-4)8(11)12;;/h1-3H,(H,13,14,15);2*1H2

InChI key

NONUFBPOKYHACM-UHFFFAOYSA-N

Application

The biotin end of photobiotin-labeled DNA is modified with a hapten derived from this product.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G H Keller et al.
Analytical biochemistry, 177(2), 392-395 (1989-03-01)
A photoactivatable reagent for introducing haptens onto DNA probes has been prepared using a commercially available bifunctional linker arm reagent and amino-derivatized 2,4-dinitrophenyl (DNP). The resulting compound (photo-DNP) couples efficiently to DNA using an ordinary sunlamp. Under optimum conditions, about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service