Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380296

Sigma-Aldrich

Yttrium(III) isopropoxide solution

20-25 wt. % in toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(OCH(CH3)2)3
CAS Number:
Molecular Weight:
266.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: yttrium
reagent type: catalyst

concentration

20-25 wt. % in toluene

bp

106 °C

density

0.955 g/mL at 25 °C

SMILES string

CC(C)O[Y](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Y/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

PYLIDHFYDYRZSC-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service