Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

379395

Sigma-Aldrich

Ethyl 1-piperidineacetate

98%

Synonym(s):

Piperidin-1-ylacetic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

109-111 °C/25 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CN1CCCCC1

InChI

1S/C9H17NO2/c1-2-12-9(11)8-10-6-4-3-5-7-10/h2-8H2,1H3

InChI key

LXZHLNWWDDNIOJ-UHFFFAOYSA-N

General description

Ethyl 1-piperidineacetate (Piperidin-1-ylacetic acid ethyl ester) is an α-amino acid ethyl ester derivative. Thermal decomposition of ethyl 1-piperidineacetate in gas phase has been studied by ab initio theoretical methods.[1] The kinetics of gas-phase decomposition of ethyl 1-piperidineacetate to the corresponding α-amino acid derivative and ethylene over the temperature range of 360-430°C and pressure range of 26-86Torr has been investigated.[2] Alkylation reaction of ethyl 1-piperidineacetate with ethyl bromoacetate in absolute ethanol has been reported to obey second order kinetics.[3]

Application

Reactant for synthesis of:
Nicotinic acid receptor agonists for treatment of dyslipidemia
1-Alkylcyclanols with side chain functions
Benzhydrylamides for pharmacochemical studies

Reactant for:
Elimination reactions
Hydrogenation of esters to alcohols

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of neutral amino acid decomposition in the gas phase. The elimination kinetics of N, N-dimethylglycine ethyl ester, ethyl 1-piperidineacetate, and N, N-dimethylglycine.
Ensuncho A, et al.
International Journal of Chemical Kinetics, 33(8), 465-471 (2001)
Theoretical study of the gas-phase decomposition of neutral α-amino acid ethyl esters. Part 2-Elimination of ethyl picolinate and ethyl 1-methylpipecolinate.
Notario R, et al.
Journal of Physical Organic Chemistry, 16(3), 166-174 (2003)
Alkylation of N-carbethoxy tertiary amines with ethyl bromoacetate.
Duty RC and Gurnea RL.
The Journal of Organic Chemistry, 35(6), 1800-1802 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service