About This Item
assay
95%
mp
232-235 °C (lit.)
SMILES string
c1ccc-2c(c1)-c3ccccc3-c4ccccc4-c5ccccc-25
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service