Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

378607

Sigma-Aldrich

2-(4-Methoxybenzyloxycarbonyloxyimino)-2-phenylacetonitrile

98%

Synonym(s):

MOZ-ON

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2OCO2N=C(C6H5)CN
CAS Number:
Molecular Weight:
310.30
Beilstein/REAXYS Number:
3006152
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

110-115 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(COC(=O)O\N=C(\C#N)c2ccccc2)cc1

InChI

1S/C17H14N2O4/c1-21-15-9-7-13(8-10-15)12-22-17(20)23-19-16(11-18)14-5-3-2-4-6-14/h2-10H,12H2,1H3/b19-16-

InChI key

IPRBOQUKBZNCAG-MNDPQUGUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(4-Methoxybenzyloxycarbonyloxyimino)-2-phenylacetonitrile (MOZ-ON) is an organic building block.

Application

2-(4-Methoxybenzyloxycarbonyloxyimino)-2-phenylacetonitrile (MOZ-ON) is suitable for use in the synthesis of biotinylated ampicillin sulfone, a bifunctional mechanism-based inhibitor for the selection of active site serine β-lactamases.[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Amstutz et al.
Journal of the American Chemical Society, 124(32), 9396-9403 (2002-08-09)
We report what is, to our knowledge, the first in vitro selection for catalytic activity based on catalytic turnover by using ribosome display, a method which does not involve living cells at any step. RTEM-beta-lactamase was functionally displayed on ribosomes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service