Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

377058

Sigma-Aldrich

Poly[4-vinylpyridinium poly(hydrogen fluoride)]

Synonym(s):

PVPHF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

InChI

1S/C21H23N3.3FH/c1-17(19-6-12-23-13-7-19)16-21(20-8-14-24-15-9-20)3-2-18-4-10-22-11-5-18;;;/h4-15,17,21H,2-3,16H2,1H3;3*1H

InChI key

VXDLBAQFZXDLIF-UHFFFAOYSA-N

Application

Convenient source of HF for fluorinations.[1]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 267-267 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service