Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376957

tert-Butyl nitrite

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CONO
CAS Number:
Molecular Weight:
103.12
Beilstein/REAXYS Number:
1209339
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

96%

form

liquid

refractive index

n20/D 1.368 (lit.)

bp

61-63 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)ON=O

InChI

1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3

InChI key

IOGXOCVLYRDXLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

14.0 °F - closed cup

flash_point_c

-10 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga A Guselnikova et al.
Beilstein journal of nanotechnology, 6, 1192-1198 (2015-07-15)
A novel approach for the in situ synthesis of zerovalent aryl-coated iron nanoparticles (NPs) based on diazonium salt chemistry is proposed. Surface-modified zerovalent iron NPs (ZVI NPs) were prepared by simple chemical reduction of iron(III) chloride aqueous solution followed by
Michael Lesslie et al.
European journal of mass spectrometry (Chichester, England), 21(3), 589-597 (2015-08-27)
Radical migration, both intramolecular and intermolecular, from the tyrosine phenoxyl radical Tyr(O(∙)) to the cysteine radical Cys(S(∙)) in model peptide systems was observed in the gas phase. Ion-molecule reactions (IMRs) between the radical cation of homotyrosine and propyl thiol resulted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service