Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376647

2-Chloro-3,5-difluoroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClF2N
CAS Number:
Molecular Weight:
163.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

SMILES string

Nc1cc(F)cc(F)c1Cl

InChI

1S/C6H4ClF2N/c7-6-4(9)1-3(8)2-5(6)10/h1-2H,10H2

InChI key

XDHOQZCRADUDRI-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service